Assay Details
Gene Symbol: LOC101715566 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848494.3 | XP_004848551.1 | 7 - 8 | 1206 | 75 | |
XM_021261384.1 | XP_021117043.1 | 12 - 13 | 1957 | 75 | ||
XM_021261385.1 | XP_021117044.1 | 13 - 14 | 1752 | 75 | ||
XM_021261386.1 | XP_021117045.1 | 12 - 13 | 1822 | 75 | ||
XM_021261387.1 | XP_021117046.1 | 8 - 9 | 1085 | 75 | ||
XM_021261388.1 | XP_021117047.1 | 7 - 8 | 976 | 75 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
