Assay Details
Gene Symbol: Pou6f1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004862946.2 | XP_004863003.1 | 2 - 3 | 458 | 61 | |
XM_004862947.3 | XP_004863004.1 | 2 - 3 | 458 | 61 | ||
XM_013079056.2 | XP_012934510.1 | 2 - 3 | 459 | 61 | ||
XM_021243065.1 | XP_021098724.1 | 2 - 3 | 394 | 61 | ||
XM_021243066.1 | XP_021098725.1 | 2 - 3 | 464 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
