Assay Details
Gene Symbol: LOC101714324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068852.2 | XP_012924306.1 | 3 - 4 | 469 | 61 | |
XM_021249578.1 | XP_021105237.1 | 3 - 4 | 469 | 61 | ||
XM_021249580.1 | XP_021105239.1 | 3 - 4 | 470 | 61 | ||
XM_021249582.1 | XP_021105241.1 | 3 - 4 | 468 | 61 | ||
XM_021249588.1 | XP_021105247.1 | 3 - 4 | 468 | 61 | ||
XM_021249597.1 | XP_021105256.1 | 3 - 4 | 468 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
