Assay Details
Gene Symbol: LOC101714324 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013068852.2 | XP_012924306.1 | 8 - 9 | 1011 | 100 | |
XM_021249578.1 | XP_021105237.1 | 8 - 9 | 1011 | 100 | ||
XM_021249580.1 | XP_021105239.1 | 8 - 9 | 1012 | 100 | ||
XM_021249588.1 | XP_021105247.1 | 8 - 9 | 1010 | 100 | ||
XM_021249597.1 | XP_021105256.1 | 8 - 9 | 1010 | 100 |
