Assay Details
Gene Symbol: CUNH17orf80 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013078168.2 | XP_012933622.1 | 2 - 3 | 218 | 82 | |
XM_013078169.2 | XP_012933623.1 | 2 - 3 | 271 | 82 | ||
XM_021240995.1 | XP_021096654.1 | 2 - 3 | 107 | 82 | ||
XM_021240996.1 | XP_021096655.1 | 2 - 3 | 218 | 82 | ||
XM_021240997.1 | XP_021096656.1 | 2 - 3 | 218 | 82 | ||
XM_021240998.1 | XP_021096657.1 | 2 - 3 | 219 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013078168.2, XM_013078169.2, XM_021240995.1, XM_021240996.1, XM_021240997.1, XM_021240998.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
