Assay Details
Gene Symbol: Cnnm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245641.1 | XP_021101300.1 | 5 - 6 | 2245 | 63 | |
XM_021245642.1 | XP_021101301.1 | 5 - 6 | 2245 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() |
