Assay Details
Gene Symbol: Dio2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004837107.3 | XP_004837164.2 | 1 - 2 | 1152 | 80 | |
XM_013066956.2 | XP_012922410.1 | 1 - 2 | 1154 | 80 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
