Assay Details
Gene Symbol: Smim5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004860691.3 | XP_004860748.1 | 2 - 3 | 663 | 66 | |
XM_021241220.1 | XP_021096879.1 | 2 - 3 | 323 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
