Assay Details
Gene Symbol: Tnfsf13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013076791.2 | XP_012932245.1 | 5 - 6 | 948 | 65 | |
XM_021238143.1 | XP_021093802.1 | 5 - 6 | 978 | 65 | ||
XM_021238145.1 | XP_021093804.1 | 7 - 8 | 876 | 65 | ||
XM_021238146.1 | XP_021093805.1 | 4 - 5 | 900 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
