Assay Details
| Gene Symbol: LOC106007555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248862.1 | XP_021104521.1 | 4 - 5 | 1081 | 100 | |
More Information
Set Membership: | 
		   
					
						
							 | 
				
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC106007555| Gene Symbol: LOC106007555 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_021248862.1 | XP_021104521.1 | 4 - 5 | 1081 | 100 | |
Set Membership: | 
		   
					
						
							 |