Assay Details
| Gene Symbol: Znf316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004840429.3 | XP_004840486.1 | 6 - 7 | 1007 | 93 | |
| XM_013070620.2 | XP_012926074.1 | 6 - 7 | 1046 | 93 | ||
More Information
Set Membership: |
|

Znf316
Genomic Map