Assay Details
Gene Symbol: Mbd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004855174.3 | XP_004855231.1 | 1 - 2 | 695 | 63 | |
XM_021236639.1 | XP_021092298.1 | 1 - 2 | 697 | 63 | ||
XM_021236640.1 | XP_021092299.1 | 1 - 2 | 730 | 63 | ||
XM_021236641.1 | XP_021092300.1 | 1 - 2 | 729 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
