Assay Details
| Gene Symbol: LOC101707650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859395.2 | XP_004859452.1 | 1 - 2 | 587 | 75 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC101707650| Gene Symbol: LOC101707650 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004859395.2 | XP_004859452.1 | 1 - 2 | 587 | 75 | |