Assay Details
| Gene Symbol: LOC101702503 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004864212.2 | XP_004864269.1 | 2 - 3 | 209 | 78 | |
| XM_021243911.1 | XP_021099570.1 | 2 - 3 | 203 | 78 | ||
| XM_021243912.1 | XP_021099571.1 | 2 - 3 | 203 | 78 | ||
More Information
Set Membership: |
|

LOC101702503
Genomic Map