Assay Details
Gene Symbol: Tedc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004864646.2 | XP_004864703.1 | 7 - 8 | 983 | 58 | |
XM_021244358.1 | XP_021100017.1 | 7 - 8 | 1065 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
