Assay Details
| Gene Symbol: Slc1a7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004869226.3 | XP_004869283.1 | 7 - 8 | 1147 | 79 | |
| XM_021247996.1 | XP_021103655.1 | 9 - 10 | 6405 | 79 | ||
| XM_021247997.1 | XP_021103656.1 | 9 - 10 | 5504 | 79 | ||
| XM_021247998.1 | XP_021103657.1 | 8 - 9 | 5399 | 79 | ||
| XM_021247999.1 | XP_021103658.1 | 8 - 9 | 4347 | 79 | ||
| XM_021248000.1 | XP_021103659.1 | 8 - 9 | 1297 | 79 | ||
| XM_021248001.1 | XP_021103660.1 | 8 - 9 | 7389 | 79 | ||
| XM_021248002.1 | XP_021103661.1 | 7 - 8 | 7263 | 79 | ||
| XM_021248004.1 | XP_021103663.1 | 8 - 9 | 7389 | 79 | ||
| XM_021248005.1 | XP_021103664.1 | 8 - 9 | 7389 | 79 | ||
More Information
Set Membership: |
|

Slc1a7
Genomic Map