Assay Details
| Gene Symbol: Sirpa | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_013070682.2 | XP_012926136.1 | 7 - 8 | 1311 | 65 | |
| XM_021254966.1 | XP_021110625.1 | 6 - 7 | 1460 | 65 | ||
| XM_021254967.1 | XP_021110626.1 | 6 - 7 | 1460 | 65 | ||
| XM_021254968.1 | XP_021110627.1 | 4 - 5 | 809 | 65 | ||
| XM_021254969.1 | XP_021110628.1 | 4 - 5 | 809 | 65 | ||
More Information
Set Membership: |
|

Sirpa
Genomic Map