Assay Details
| Gene Symbol: Bicral | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846563.2 | XP_004846620.1 | 6 - 7 | 1970 | 71 | |
| XM_021259801.1 | XP_021115460.1 | 6 - 7 | 1969 | 71 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Bicral| Gene Symbol: Bicral | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004846563.2 | XP_004846620.1 | 6 - 7 | 1970 | 71 | |
| XM_021259801.1 | XP_021115460.1 | 6 - 7 | 1969 | 71 | ||