Assay Details
Gene Symbol: LOC101702087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853091.2 | XP_004853148.2 | 15 - 16 | 2523 | 138 | |
XM_021264913.1 | XP_021120572.1 | 15 - 16 | 2523 | 138 | ||
XM_021264914.1 | XP_021120573.1 | 15 - 16 | 2591 | 138 | ||
XM_021264915.1 | XP_021120574.1 | 15 - 16 | 2524 | 138 |

More Information
Set Membership: |
![]() ![]() |
