Assay Details
Gene Symbol: LOC101702087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853091.2 | XP_004853148.2 | 27 - 28 | 4082 | 76 | |
XM_021264913.1 | XP_021120572.1 | 27 - 28 | 4106 | 76 | ||
XM_021264914.1 | XP_021120573.1 | 27 - 28 | 4174 | 76 | ||
XM_021264915.1 | XP_021120574.1 | 27 - 28 | 4107 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() |
