Assay Details
Gene Symbol: LOC101702087 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004853091.2 | XP_004853148.2 | 6 - 7 | 1107 | 97 | |
XM_021264913.1 | XP_021120572.1 | 6 - 7 | 1107 | 97 | ||
XM_021264914.1 | XP_021120573.1 | 6 - 7 | 1175 | 97 | ||
XM_021264915.1 | XP_021120574.1 | 6 - 7 | 1108 | 97 |
