Assay Details
Gene Symbol: Apc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004865947.2 | XP_004866004.1 | 7 - 8 | 829 | 56 | |
XM_004865948.2 | XP_004866005.1 | 7 - 8 | 826 | 56 | ||
XM_004865949.3 | XP_004866006.1 | 7 - 8 | 758 | 56 | ||
XM_021244848.1 | XP_021100507.1 | 7 - 8 | 983 | 56 |

More Information
Set Membership: |
![]() ![]() ![]() |
