Assay Details
Gene Symbol: Was | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871944.3 | XP_004872001.1 | 3 - 4 | 1250 | 63 | |
XM_004871947.2 | XP_004872004.1 | 4 - 5 | 913 | 63 | ||
XM_004871948.2 | XP_004872005.1 | 4 - 5 | 449 | 63 | ||
XM_004871949.2 | XP_004872006.1 | 3 - 4 | 388 | 63 | ||
XM_013068213.2 | XP_012923667.1 | 3 - 4 | 833 | 63 | ||
XM_021249978.1 | XP_021105637.1 | 5 - 6 | 650 | 63 | ||
XM_021249979.1 | XP_021105638.1 | 5 - 6 | 652 | 63 | ||
XM_021249980.1 | XP_021105639.1 | 5 - 6 | 537 | 63 | ||
XM_021249981.1 | XP_021105640.1 | 5 - 6 | 547 | 63 | ||
XM_021249982.1 | XP_021105641.1 | 4 - 5 | 478 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
