Assay Details
Gene Symbol: Spaca6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013066323.2 | XP_012921777.1 | 6 - 7 | 741 | 74 | |
XM_021246273.1 | XP_021101932.1 | 6 - 7 | 774 | 74 | ||
XM_021246274.1 | XP_021101933.1 | 6 - 7 | 736 | 74 | ||
XM_021246275.1 | XP_021101934.1 | 6 - 7 | 693 | 74 | ||
XM_021246276.1 | XP_021101935.1 | 6 - 7 | 774 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
