Assay Details
Gene Symbol: Ccdc82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004838081.3 | XP_004838138.1 | 8 - 9 | 1970 | 85 | |
XM_004838083.3 | XP_004838140.1 | 8 - 9 | 1586 | 85 | ||
XM_013069646.2 | XP_012925100.1 | 9 - 10 | 1622 | 85 | ||
XM_021252685.1 | XP_021108344.1 | 8 - 9 | 1666 | 85 | ||
XM_021252686.1 | XP_021108345.1 | 8 - 9 | 1506 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
