Assay Details
Gene Symbol: Klf12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845251.2 | XP_004845308.1 | 5 - 6 | 1090 | 65 | |
XM_004845252.2 | XP_004845309.1 | 6 - 7 | 1067 | 65 | ||
XM_004845253.2 | XP_004845310.1 | 5 - 6 | 1006 | 65 | ||
XM_021258671.1 | XP_021114330.1 | 6 - 7 | 1133 | 65 | ||
XM_021258672.1 | XP_021114331.1 | 3 - 4 | 706 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
