Assay Details
Gene Symbol: Kctd20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004846685.2 | XP_004846742.1 | 7 - 8 | 1108 | 76 | |
XM_004846686.2 | XP_004846743.1 | 7 - 8 | 1220 | 76 | ||
XM_013072985.2 | XP_012928439.1 | 5 - 6 | 711 | 76 | ||
XM_021259920.1 | XP_021115579.1 | 6 - 7 | 910 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
