Assay Details
Gene Symbol: Mettl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004857642.3 | XP_004857699.1 | 2 - 3 | 723 | 82 | |
XM_021238580.1 | XP_021094239.1 | 2 - 3 | 740 | 82 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
