Assay Details
| Gene Symbol: Bhmt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004870202.3 | XP_004870259.1 | 3 - 4 | 355 | 60 | |
| XM_013067596.1 | XP_012923050.1 | 4 - 5 | 410 | 60 | ||
More Information
Set Membership: |
|

Bhmt2
Genomic Map