Assay Details
Gene Symbol: Znf684 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004871781.2 | XP_004871838.1 | 4 - 5 | 503 | 78 | |
XM_021249938.1 | XP_021105597.1 | 5 - 6 | 671 | 78 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
