Assay Details
Gene Symbol: LOC101712837 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021245921.1 | XP_021101580.1 | 4 - 5 | 974 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
