Assay Details
Gene Symbol: LOC101699552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004847776.3 | XP_004847833.1 | 4 - 5 | 617 | 94 | |
XM_004847777.3 | XP_004847834.1 | 4 - 5 | 581 | 94 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
