Assay Details
| Gene Symbol: Nadk2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004848612.2 | XP_004848669.1 | 3 - 4 | 468 | 67 | |
| XM_004848613.2 | XP_004848670.1 | 3 - 4 | 468 | 67 | ||
| XM_013073513.2 | XP_012928967.1 | 3 - 4 | 274 | 67 | ||
| XM_021261495.1 | XP_021117154.1 | 3 - 4 | 468 | 67 | ||
| XM_021261496.1 | XP_021117155.1 | 4 - 5 | 364 | 67 | ||
| XM_021261497.1 | XP_021117156.1 | 5 - 6 | 434 | 67 | ||
| XM_021261498.1 | XP_021117157.1 | 4 - 5 | 369 | 67 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_013073513.2, XM_021261496.1, XM_021261497.1, XM_021261498.1 transcripts.
Set Membership: |
|

Nadk2
Genomic Map