Assay Details
| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 13 - 14 | 3617 | 76 | |
| XM_021262644.1 | XP_021118303.1 | 13 - 14 | 3619 | 76 | ||
| XM_021262645.1 | XP_021118304.1 | 12 - 13 | 3293 | 76 | ||
| XM_021262646.1 | XP_021118305.1 | 10 - 11 | 3118 | 76 | ||
| XM_021262647.1 | XP_021118306.1 | 13 - 14 | 3622 | 76 | ||
| XM_021262648.1 | XP_021118307.1 | 7 - 8 | 2619 | 76 | ||

Virma
Genomic Map