Assay Details
| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 22 - 23 | 5377 | 90 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Virma| Gene Symbol: Virma | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850114.3 | XP_004850171.1 | 22 - 23 | 5377 | 90 | |