Assay Details
| Gene Symbol: Aunip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004850615.2 | XP_004850672.1 | 2 - 3 | 1069 | 66 | |
| XM_004850617.3 | XP_004850674.1 | 3 - 4 | 1193 | 66 | ||
| XM_021263090.1 | XP_021118749.1 | 2 - 3 | 198 | 66 | ||
| XM_021263092.1 | XP_021118751.1 | 2 - 3 | 352 | 66 | ||
More Information
Set Membership: |
|

Aunip
Genomic Map