Assay Details
| Gene Symbol: Tmcc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004862111.3 | XP_004862168.1 | 4 - 5 | 2253 | 87 | |
| XM_004862112.3 | XP_004862169.1 | 5 - 6 | 1895 | 87 | ||
| XM_004862114.2 | XP_004862171.1 | 4 - 5 | 2140 | 87 | ||
| XM_004862115.2 | XP_004862172.1 | 4 - 5 | 1787 | 87 | ||
| XM_013078652.2 | XP_012934106.1 | 3 - 4 | 1290 | 87 | ||
| XM_013078653.2 | XP_012934107.1 | 4 - 5 | 1358 | 87 | ||
| XM_021242375.1 | XP_021098034.1 | 5 - 6 | 2016 | 87 | ||
More Information
Set Membership: |
|

Tmcc2
Genomic Map