Assay Details
| Gene Symbol: Tbxa2r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_004865845.2 | XP_004865902.1 | 2 - 3 | 1176 | 104 | |
| XM_021245146.1 | XP_021100805.1 | 3 - 4 | 1216 | 104 | ||
| XM_021245147.1 | XP_021100806.1 | 2 - 3 | 2110 | 104 | ||
More Information
Set Membership: |
|

Tbxa2r
Genomic Map