Assay Details
Gene Symbol: Znf865 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004866630.3 | XP_004866687.1 | 1 - 2 | 1262 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
