Assay Details
Gene Symbol: Mtg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_013070793.2 | XP_012926247.1 | 2 - 3 | 619 | 65 | |
XM_021255206.1 | XP_021110865.1 | 2 - 3 | 364 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
