Assay Details
Gene Symbol: Tmem91 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021250943.1 | XP_021106602.1 | 3 - 4 | 604 | 79 | |
XM_021250944.1 | XP_021106603.1 | 3 - 4 | 565 | 79 | ||
XM_021250945.1 | XP_021106604.1 | 3 - 4 | 521 | 79 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
