Assay Details
Gene Symbol: Eid1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004835441.2 | XP_004835498.1 | 1 | 121 | 61 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_004835441.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
