Assay Details
Gene Symbol: Olr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004845889.2 | XP_004845946.1 | 2 - 3 | 298 | 70 | |
XM_021259179.1 | XP_021114838.1 | 2 - 3 | 333 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
