Assay Details
Gene Symbol: LOC101723225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848054.3 | XP_004848111.1 | 12 - 13 | 1698 | 109 | |
XM_013073350.2 | XP_012928804.1 | 12 - 13 | 1700 | 109 | ||
XM_013073351.2 | XP_012928805.1 | 12 - 13 | 1700 | 109 | ||
XM_021261057.1 | XP_021116716.1 | 12 - 13 | 2048 | 109 | ||
XM_021261058.1 | XP_021116717.1 | 12 - 13 | 1700 | 109 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
