Assay Details
Gene Symbol: LOC101723225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004848054.3 | XP_004848111.1 | 1 - 2 | 181 | 81 | |
XM_013073350.2 | XP_012928804.1 | 1 - 2 | 183 | 81 | ||
XM_013073351.2 | XP_012928805.1 | 1 - 2 | 183 | 81 | ||
XM_021261058.1 | XP_021116717.1 | 1 - 2 | 183 | 81 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
