Assay Details
Gene Symbol: LOC101723006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851264.3 | XP_004851321.1 | 3 - 4 | 428 | 116 | |
XM_004851266.3 | XP_004851323.1 | 4 - 5 | 1686 | 116 | ||
XM_004851267.3 | XP_004851324.1 | 4 - 5 | 324 | 116 | ||
XM_004851269.3 | XP_004851326.1 | 3 - 4 | 567 | 116 | ||
XM_021263677.1 | XP_021119336.1 | 5 - 6 | 411 | 116 | ||
XM_021263678.1 | XP_021119337.1 | 5 - 6 | 414 | 116 | ||
XM_021263679.1 | XP_021119338.1 | 6 - 7 | 487 | 116 | ||
XM_021263680.1 | XP_021119339.1 | 4 - 5 | 326 | 116 | ||
XM_021263681.1 | XP_021119340.1 | 5 - 6 | 418 | 116 | ||
XM_021263682.1 | XP_021119341.1 | 4 - 5 | 2729 | 116 | ||
XM_021263683.1 | XP_021119342.1 | 4 - 5 | 1380 | 116 | ||
XM_021263684.1 | XP_021119343.1 | 4 - 5 | 1492 | 116 | ||
XM_021263685.1 | XP_021119344.1 | 3 - 4 | 211 | 116 | ||
XM_021263686.1 | XP_021119345.1 | 5 - 6 | 453 | 116 |

More Information
Set Membership: |
![]() ![]() |
