Assay Details
Gene Symbol: LOC101723006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851264.3 | XP_004851321.1 | 5 - 6 | 684 | 70 | |
XM_004851266.3 | XP_004851323.1 | 6 - 7 | 1942 | 70 | ||
XM_004851267.3 | XP_004851324.1 | 6 - 7 | 580 | 70 | ||
XM_004851269.3 | XP_004851326.1 | 5 - 6 | 823 | 70 | ||
XM_021263677.1 | XP_021119336.1 | 7 - 8 | 667 | 70 | ||
XM_021263678.1 | XP_021119337.1 | 7 - 8 | 670 | 70 | ||
XM_021263679.1 | XP_021119338.1 | 8 - 9 | 743 | 70 | ||
XM_021263680.1 | XP_021119339.1 | 6 - 7 | 582 | 70 | ||
XM_021263681.1 | XP_021119340.1 | 7 - 8 | 674 | 70 | ||
XM_021263682.1 | XP_021119341.1 | 6 - 7 | 2985 | 70 | ||
XM_021263683.1 | XP_021119342.1 | 6 - 7 | 1636 | 70 | ||
XM_021263684.1 | XP_021119343.1 | 6 - 7 | 1748 | 70 | ||
XM_021263685.1 | XP_021119344.1 | 5 - 6 | 467 | 70 | ||
XM_021263686.1 | XP_021119345.1 | 7 - 8 | 709 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
