Assay Details
Gene Symbol: LOC101723006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851264.3 | XP_004851321.1 | 6 - 7 | 864 | 91 | |
XM_004851266.3 | XP_004851323.1 | 7 - 8 | 2122 | 91 | ||
XM_004851267.3 | XP_004851324.1 | 7 - 8 | 760 | 91 | ||
XM_004851269.3 | XP_004851326.1 | 6 - 7 | 1003 | 91 | ||
XM_021263677.1 | XP_021119336.1 | 8 - 9 | 847 | 91 | ||
XM_021263678.1 | XP_021119337.1 | 8 - 9 | 850 | 91 | ||
XM_021263679.1 | XP_021119338.1 | 9 - 10 | 923 | 91 | ||
XM_021263680.1 | XP_021119339.1 | 7 - 8 | 762 | 91 | ||
XM_021263681.1 | XP_021119340.1 | 8 - 9 | 854 | 91 | ||
XM_021263682.1 | XP_021119341.1 | 7 - 8 | 3165 | 91 | ||
XM_021263683.1 | XP_021119342.1 | 7 - 8 | 1816 | 91 | ||
XM_021263684.1 | XP_021119343.1 | 7 - 8 | 1928 | 91 | ||
XM_021263685.1 | XP_021119344.1 | 6 - 7 | 647 | 91 | ||
XM_021263686.1 | XP_021119345.1 | 8 - 9 | 889 | 91 |

More Information
Set Membership: |
![]() ![]() ![]() |
