Assay Details
Gene Symbol: LOC101723006 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851264.3 | XP_004851321.1 | 8 - 9 | 988 | 92 | |
XM_004851266.3 | XP_004851323.1 | 9 - 10 | 2246 | 92 | ||
XM_004851267.3 | XP_004851324.1 | 9 - 10 | 884 | 92 | ||
XM_021263679.1 | XP_021119338.1 | 11 - 12 | 1047 | 92 | ||
XM_021263681.1 | XP_021119340.1 | 10 - 11 | 978 | 92 | ||
XM_021263682.1 | XP_021119341.1 | 9 - 10 | 3289 | 92 | ||
XM_021263683.1 | XP_021119342.1 | 9 - 10 | 1940 | 92 | ||
XM_021263684.1 | XP_021119343.1 | 9 - 10 | 2052 | 92 | ||
XM_021263685.1 | XP_021119344.1 | 8 - 9 | 771 | 92 | ||
XM_021263686.1 | XP_021119345.1 | 10 - 11 | 1013 | 92 |
