Assay Details
Gene Symbol: Adck2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_004851358.2 | XP_004851415.1 | 4 - 5 | 1786 | 59 | |
XM_021263701.1 | XP_021119360.1 | 4 - 5 | 1790 | 59 | ||
XM_021263702.1 | XP_021119361.1 | 4 - 5 | 1790 | 59 | ||
XM_021263703.1 | XP_021119362.1 | 4 - 5 | 1793 | 59 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
